Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016706.2 | NP_057915.2 | 2 | 1110 | |
GenBank mRNA | AF208663.1 | - | 2 | 1092 | |
AK146366.1 | - | 2 | 1094 | ||
BC031167.1 | - | 2 | 1125 | 4983919 | |
CF948658.1 | - | 1 | 668 | ||
CF949734.1 | - | 1 | 384 |