Gene Transcripts

Gene Symbol
Csf2
Entrez Gene ID
12981
Gene Name
colony stimulating factor 2 (granulocyte-macrophage)
Gene Aliases
Csfgm, GMCSF, Gm-CSf, MGI-IGM
Chromosome Location
Chr. 11: 54247270 - 54249899 on Build GRCm38
UniGene ID
Mm.4922
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009969.4 NP_034099.2 4 642
GenBank mRNA AK053196.1 - 4 642
BC116878.1 - 4 376 40126197
BC116880.1 - 4 376 40126199
EU366957.1 - 4 353
M11848.1 - 4 311
X02333.1 - 4 526
X03019.1 - 4 385
X03221.1 - 4 384
X05906.1 - 4 284

View Full Product Details