Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007807.5 | NP_031833.3 | 6 | 678 | |
GenBank mRNA | AK033570.1 | - | 6 | 622 | |
AK053920.1 | - | 6 | 622 | ||
AK152257.1 | - | 6 | 649 | ||
AK153154.1 | - | 6 | 649 | ||
AK153259.1 | - | 6 | 649 | ||
AK154126.1 | - | 6 | 678 | ||
AK160031.1 | - | 1 | 80 | ||
AK171526.1 | - | 6 | 664 | ||
BC042838.1 | - | 6 | 645 | ||
BC071229.1 | - | 6 | 633 | 4979804 | |
FJ168469.1 | - | 6 | 667 | ||
U43384.1 | - | 6 | 604 |