Gene Transcripts

Gene Symbol
Dbx1
Entrez Gene ID
13172
Gene Name
developing brain homeobox 1
Gene Aliases
AI426026, Dbx
Chromosome Location
Chr. 7: 49631499 - 49636835 on Build GRCm38
UniGene ID
Mm.307176
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005232.1 NP_001005232.1 1 249
GenBank mRNA BC082541.1 - 1 249
S75837.1 - 1 262

View Full Product Details