Gene Transcripts

Gene Symbol
Dll3
Entrez Gene ID
13389
Gene Name
delta-like 3 (Drosophila)
Gene Aliases
pu, pudgy
Chromosome Location
Chr. 7: 28293555 - 28301785 on Build GRCm38
UniGene ID
Mm.12896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007866.2 NP_031892.2 7 1675
GenBank mRNA AB013440.1 - 7 1662
AK144282.1 - 7 1676
BC052002.1 - 7 1652 6404029

View Full Product Details