Gene Transcripts

Gene Symbol
Dlx3
Entrez Gene ID
13393
Gene Name
distal-less homeobox 3
Gene Aliases
AV237891, Dlx-3
Chromosome Location
Chr. 11: 95120117 - 95125293 on Build GRCm38
UniGene ID
Mm.5194
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010055.3 NP_034185.1 3 750
GenBank mRNA BC058852.1 - 3 748
S74448.1 - 2 137
S81932.1 - 3 545
U79738.1 - 3 377

View Full Product Details