Gene Transcripts
- Gene Symbol
- Tmc1
- Entrez Gene ID
- 13409
- Gene Name
- transmembrane channel-like gene family 1
- Gene Aliases
- 4933416G09Rik, Beethoven, Bth, CWEA1, dn
- Chromosome Location
- Chr. 19: 20783458 - 21037152 on Build GRCm38
- UniGene ID
- Mm.219585
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028953.2 |
NP_083229.1
|
14 |
1611 |
|
GenBank mRNA |
AF417579.1 |
- |
14 |
1611 |
|
View Full Product Details