Gene Transcripts

Gene Symbol
Fgf15
Entrez Gene ID
14170
Gene Name
fibroblast growth factor 15
Gene Aliases
FGF19
Chromosome Location
Chr. 7: 144896532 - 144900953 on Build GRCm38
UniGene ID
Mm.3904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008003.2 NP_032029.1 2 427
GenBank mRNA AF007268.1 - 2 427
AK017829.1 - 2 429
AK052093.1 - 1 613
BC021328.1 - 2 419 5066286

View Full Product Details