Gene Transcripts

Gene Symbol
Gaa
Entrez Gene ID
14387
Gene Name
glucosidase, alpha, acid
Gene Aliases
E430018M07Rik
Chromosome Location
Chr. 11: 119267967 - 119285698 on Build GRCm38
UniGene ID
Mm.4793
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159324.1 NP_001152796.1 14 2110
NM_008064.3 NP_032090.3 14 2200
GenBank mRNA AK052211.1 - 14 2181
AK088481.1 - 14 2176
AK139333.1 - 14 2148
AK141546.1 - 14 2183
AK145631.1 - 13 1616
AK146538.1 - 14 2104
AK150970.1 - 14 2158
BC010210.1 - 14 2164 3969127
U49351.1 - 14 2052

View Full Product Details