Gene Transcripts

Gene Symbol
Gm2a
Entrez Gene ID
14667
Gene Name
GM2 ganglioside activator protein
Gene Aliases
AA408702, AW215435, GM2-AP, SAP-3
Chromosome Location
Chr. 11: 55097985 - 55113029 on Build GRCm38
UniGene ID
Mm.287807
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010299.3 NP_034429.1 3, 4 614
GenBank mRNA AK149642.1 - 3, 4 507
AK152256.1 - 3, 4 506
AK210428.1 - 1, 2 191
BC004651.1 - 3, 4 482 3482848
CT010289.1 - 3, 4 435
L19526.1 - 3, 4 448
U09816.1 - 3, 4 491

View Full Product Details