Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010307.3 | NP_034437.1 | 7 | 852 | |
NM_177137.5 | NP_796111.2 | 7 | 1017 | ||
GenBank mRNA | AK158831.1 | - | 7 | 853 | |
AY179169.1 | - | 7 | 586 | ||
BC078439.1 | - | 7 | 979 | 5685967 |