Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127318.1 | NP_001120790.1 | 16 | 1889 | |
NM_145067.3 | NP_659504.2 | 15 | 1817 | ||
GenBank mRNA | AK136496.1 | - | 16 | 1913 | |
BC034064.1 | - | 9 | 799 | ||
BC099968.1 | - | 15 | 1817 | 6432585 |