Gene Transcripts

Gene Symbol
Ifi44l
Entrez Gene ID
15061
Gene Name
interferon-induced protein 44 like
Gene Aliases
H-28, H28, H28-1, NS1178
Chromosome Location
Chr. 3: 151758737 - 151762891 on Build GRCm38
UniGene ID
Mm.219544
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031367.1 NP_112735.1 805
GenBank mRNA AF336221.1 - 805

View Full Product Details