Gene Transcripts

Gene Symbol
HOXA4
Entrez Gene ID
3201
Gene Name
homeobox A4
Gene Aliases
HOX1, HOX1D
Chromosome Location
Chr. 7: 27128507 - 27130780 on Build GRCh38
UniGene ID
Hs.533357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002141.4 NP_002132.3 2 1639
GenBank mRNA AI277552.1 - 1 163

View Full Product Details