Gene Transcripts

Gene Symbol
Hoxa4
Entrez Gene ID
15401
Gene Name
homeobox A4
Gene Aliases
AV206827, Hox-1.4
Chromosome Location
Chr. 6: 52189687 - 52191703 on Build GRCm38
UniGene ID
Mm.439647
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008265.3 NP_032291.1 1, 2 536
GenBank mRNA AK076098.1 - 1, 2 537
BC107172.2 - 2, 3 243 40044111

View Full Product Details