Gene Transcripts

Gene Symbol
Htr1b
Entrez Gene ID
15551
Gene Name
5-hydroxytryptamine (serotonin) receptor 1B
Gene Aliases
Chromosome Location
Chr. 9: 81631392 - 81632552 on Build GRCm38
UniGene ID
Mm.445308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010482.1 NP_034612.1 1 458
GenBank mRNA BC119129.1 - 1 521
BC131951.1 - 1 521 40130228

View Full Product Details