Gene Transcripts

Gene Symbol
Kcnab1
Entrez Gene ID
16497
Gene Name
potassium voltage-gated channel, shaker-related subfamily, beta member 1
Gene Aliases
Akr8a8, Kvbeta1.1, mKv(beta)1
Chromosome Location
Chr. 3: 64949049 - 65378225 on Build GRCm38
UniGene ID
Mm.316402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289450.1 NP_001276379.1 12, 13 889
NM_010597.4 NP_034727.3 11, 12 1305
GenBank mRNA AF033003.1 - 11, 12 1660
AK004666.1 - 12, 13 873
AK015412.1 - 13, 14 1145
AK031362.1 - 11, 12 1306
AK138467.1 - 11, 12 1294
AK153200.1 - 12, 13 891
BC014701.1 - 11, 12 1030 4482268
U65591.1 - 11, 12 901
X97281.1 - 11, 12 1289

View Full Product Details