Gene Transcripts

Gene Symbol
Kcnc3
Entrez Gene ID
16504
Gene Name
potassium voltage gated channel, Shaw-related subfamily, member 3
Gene Aliases
KShIIID, Kcr2-3, Kv3.3
Chromosome Location
Chr. 7: 44590661 - 44604754 on Build GRCm38
UniGene ID
Mm.40312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290682.1 NP_001277611.1 2 1403
NM_008422.2 NP_032448.2 2 1403

View Full Product Details