Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133664.3 | NP_598425.2 | 3 | 1066 | |
GenBank mRNA | AK083526.1 | - | 3 | 1067 | |
BC016257.1 | - | 3 | 737 | 4912735 | |
BC025580.1 | - | 3 | 970 | 5325039 | |
BC031131.1 | - | 3 | 913 | 4954891 |