Gene Transcripts

Gene Symbol
Lum
Entrez Gene ID
17022
Gene Name
lumican
Gene Aliases
Ldc, SLRR2D
Chromosome Location
Chr. 10: 97565501 - 97572703 on Build GRCm38
UniGene ID
Mm.18888
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008524.2 NP_032550.2 2 1116
GenBank mRNA AK014312.1 - 2 812
AK075737.1 - 2 815
AK085240.2 - 2 808
AK164774.1 - 2 808
BC005550.1 - 2 794 3585672
S79461.1 - 2 797

View Full Product Details