Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039543.2 | NP_001034632.1 | 5 | 548 | |
NM_010801.3 | NP_034931.2 | 4 | 503 | ||
GenBank mRNA | AF009515.1 | - | 4 | 444 | |
AF100171.1 | - | 4 | 444 | ||
AK007292.1 | - | 4 | 502 | ||
AK133448.1 | - | 4 | 504 | ||
BC027526.1 | - | 5 | 497 | 1495721 |