Gene Transcripts

Gene Symbol
Npy1r
Entrez Gene ID
18166
Gene Name
neuropeptide Y receptor Y1
Gene Aliases
Npyr, Y1-R
Chromosome Location
Chr. 8: 66697192 - 66706798 on Build GRCm38
UniGene ID
Mm.5112
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010934.4 NP_035064.1 3 1285
GenBank mRNA AK132397.1 - 3 1286
D63818.1 - 2 1120


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BC051420.1 - 1178 6490858 Chr. - - [-]

View Full Product Details