Gene Transcripts

Gene Symbol
Nr4a2
Entrez Gene ID
18227
Gene Name
nuclear receptor subfamily 4, group A, member 2
Gene Aliases
HZF-3, NOT, Nurr1, RNR-1, TINOR, TINUR
Chromosome Location
Chr. 2: 57106830 - 57124003 on Build GRCm38
UniGene ID
Mm.3507
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139509.1 NP_001132981.1 3 1025
NM_013613.2 NP_038641.1 3 1171
GenBank mRNA AB014889.1 - 3 764
AK087495.1 - 4 1049
AK158607.1 - 3 1172
BC137715.1 - 3 1483 8860734
BC144854.1 - 3 1699
S53744.1 - 3 1062

View Full Product Details