Gene Transcripts

Gene Symbol
Pdha2
Entrez Gene ID
18598
Gene Name
pyruvate dehydrogenase E1 alpha 2
Gene Aliases
Pdhal
Chromosome Location
Chr. 3: 141210004 - 141212355 on Build GRCm38
UniGene ID
Mm.4223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008811.2 NP_032837.1 1 1510
GenBank mRNA AK006356.1 - 1 939
AK076791.1 - 1 982
BC100460.1 - 1 930 30918688
M76728.1 - 1 1507

View Full Product Details