Gene Transcripts

Gene Symbol
Pla2g2c
Entrez Gene ID
18781
Gene Name
phospholipase A2, group IIC
Gene Aliases
Chromosome Location
Chr. 4: 138725325 - 138744575 on Build GRCm38
UniGene ID
Mm.5189
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008868.3 NP_032894.2 6 1487
GenBank mRNA U18119.1 - 5 1316

View Full Product Details