Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163548.1 | NP_001157020.1 | 5 | 847 | |
NM_011182.4 | NP_035312.3 | 5 | 523 | ||
GenBank mRNA | AB013470.1 | - | 5 | 377 | |
AF084221.1 | - | 5 | 417 | ||
AK016645.1 | - | 5 | 849 | ||
AK036191.1 | - | 5 | 437 | ||
AK142407.1 | - | 5 | 512 | ||
AK159229.1 | - | 5 | 495 | ||
AK168566.1 | - | 5 | 437 | ||
AK220565.1 | - | 5 | 503 | ||
BC007189.1 | - | 4 | 264 | ||
BC035296.1 | - | 5 | 464 | 3969080 |