Gene Transcripts

Gene Symbol
Ptpn18
Entrez Gene ID
19253
Gene Name
protein tyrosine phosphatase, non-receptor type 18
Gene Aliases
FLP1, HSCF, PTP-HSCF, PTP-K1, Ptpk1
Chromosome Location
Chr. 1: 34459746 - 34473784 on Build GRCm38
UniGene ID
Mm.361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011206.2 NP_035336.2 6, 7 559
GenBank mRNA AK144353.1 - 6, 7 541
BC008512.1 - 6, 7 529 3591418
U35124.1 - 6, 7 533
U49853.1 - 6, 7 499
U52523.1 - 6, 7 547

View Full Product Details