Gene Transcripts

Gene Symbol
Ptpn20
Entrez Gene ID
19256
Gene Name
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases
typ
Chromosome Location
Chr. 14: 33589201 - 33640754 on Build GRCm38
UniGene ID
Mm.8019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008978.2 NP_033004.1 8 766
GenBank mRNA AK017003.1 - 8 829
AK029493.1 - 8 764
BC139817.1 - 7 732
D64141.1 - 7 765

View Full Product Details