Gene Transcripts

Gene Symbol
Rbl2
Entrez Gene ID
19651
Gene Name
retinoblastoma-like 2
Gene Aliases
PRB2, RBR-2, Rb2, p130
Chromosome Location
Chr. 8: 91070057 - 91123844 on Build GRCm38
UniGene ID
Mm.235580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282000.1 NP_001268929.1 4 702
NM_001282001.1 NP_001268930.1 4 702
NM_011250.4 NP_035380.3 4 702
GenBank mRNA AK155256.1 - 4 666
AK160027.1 - 4 649
BC020528.1 - 4 650 4240592
CF736271.1 - 3 330
U36799.1 - 4 602
U47333.1 - 4 644
U50850.1 - 4 610

View Full Product Details