Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166623.1 | NP_001160095.1 | 3 | 345 | |
NM_011252.4 | NP_035382.1 | 3 | 342 | ||
GenBank mRNA | AJ237846.1 | - | 3 | 272 | |
AJ237847.1 | - | 3 | 287 | ||
AK083899.1 | - | 3 | 342 | ||
AK135164.1 | - | 3 | 347 | ||
AK165006.1 | - | 3 | 288 | ||
AK207357.1 | - | 3 | 206 | ||
BC003710.1 | - | 3 | 340 | 3599678 | |
BY279114.1 | - | 3 | 342 | ||
CK619886.1 | - | 2 | 148 |