Gene Transcripts

Gene Symbol
Scn5a
Entrez Gene ID
20271
Gene Name
sodium channel, voltage-gated, type V, alpha
Gene Aliases
Nav1.5, Nav1.5c, SkM1, SkM2, mH1
Chromosome Location
Chr. 9: 119483404 - 119579030 on Build GRCm38
UniGene ID
Mm.103584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253860.1 NP_001240789.1 22 4184
NM_021544.4 NP_067519.2 23 4352
GenBank mRNA AK147254.1 - 23 4352
AK147517.1 - 23 4352

View Full Product Details