Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021882.4 | NP_068682.2 | 10, 11 | 1784 | |
GenBank mRNA | AF119092.1 | - | 5, 6 | 661 | |
AK012808.1 | - | 10, 11 | 1785 | ||
AK131956.1 | - | 10, 11 | 1786 | ||
AK186888.1 | - | 2, 3 | 305 | ||
AK191933.1 | - | 2, 3 | 305 | ||
BC075667.1 | - | 10, 11 | 1769 | ||
BC082555.1 | - | 10, 11 | 1770 | 30541702 | |
U14133.1 | - | 10, 11 | 1758 |