Gene Transcripts

Gene Symbol
Smr2
Entrez Gene ID
20600
Gene Name
submaxillary gland androgen regulated protein 2
Gene Aliases
MSG2, Vcs2
Chromosome Location
Chr. 5: 88086556 - 88109053 on Build GRCm38
UniGene ID
Mm.1397
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252679.1 NP_001239608.1 5 503
NM_001252680.1 NP_001239609.1 4 433
NM_001252681.1 NP_001239610.1 3 366
NM_021289.2 NP_067264.2 3 366
GenBank mRNA BC111829.1 - 4 396
U82376.1 - 4 433
U82377.1 - 3 366
U82378.1 - 3 366
U82379.1 - 5 503

View Full Product Details