Gene Transcripts

Gene Symbol
Serpina3m
Entrez Gene ID
20717
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 3M
Gene Aliases
3e46, AI195004, MMCM7, MMSPi2.4, Spi-2l, Spi-2rs1, Spi2-rs1, Spi2.4, spi2
Chromosome Location
Chr. 12: 104387157 - 104394259 on Build GRCm38
UniGene ID
Mm.395085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009253.2 NP_033279.2 160
GenBank mRNA AI286407.1 - 160
BC011158.1 - 162 4194074
BC053337.1 - 138 4208766
X55148.1 - 132

View Full Product Details