Gene Transcripts

Gene Symbol
Sephs2
Entrez Gene ID
20768
Gene Name
selenophosphate synthetase 2
Gene Aliases
Sps2, Ysg3
Chromosome Location
Chr. 7: 127271879 - 127274059 on Build GRCm38
UniGene ID
Mm.20294
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009266.3 NP_033292.2 1 901
GenBank mRNA AK077573.1 - 1 902
AK149516.1 - 1 898
AK168722.1 - 1 902
AK168965.1 - 1 902
BC003760.1 - 1 888
BC016508.1 - 1 893 4911695
BC028966.1 - 1 878 5374184
BC043334.1 - 1 882 5100207
U43285.1 - 1 898

View Full Product Details