Gene Transcripts

Gene Symbol
Syn1
Entrez Gene ID
20964
Gene Name
synapsin I
Gene Aliases
Syn-1, Syn1-S
Chromosome Location
Chr. X: 20860511 - 20920918 on Build GRCm38
UniGene ID
Mm.439844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110780.1 NP_001104250.1 9 1271
NM_013680.4 NP_038708.3 9 1271
GenBank mRNA BC022954.1 - 9 1249

View Full Product Details