Gene Transcripts

Gene Symbol
Tcap
Entrez Gene ID
21393
Gene Name
titin-cap
Gene Aliases
AI503981, AJ223855
Chromosome Location
Chr. 11: 98383811 - 98384953 on Build GRCm38
UniGene ID
Mm.10762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011540.2 NP_035670.2 2 840
GenBank mRNA AJ223854.1 - 2 840
AK010167.1 - 2 828
Y15845.1 - 2 830

View Full Product Details