Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166584.1 | NP_001160056.1 | 8 | 1020 | |
NM_001166585.1 | NP_001160057.1 | 5 | 538 | ||
NM_009346.3 | NP_033372.1 | 7 | 957 | ||
GenBank mRNA | AK140307.1 | - | 8 | 1021 | |
AK148375.1 | - | 7 | 923 | ||
BC052021.1 | - | 5 | 479 | ||
BC060138.1 | - | 5 | 479 | 6410573 | |
L06865.1 | - | 5 | 484 | ||
L13853.1 | - | 7 | 684 | ||
S74227.1 | - | 6 | 945 |