Gene Transcripts

Gene Symbol
Adad1
Entrez Gene ID
21744
Gene Name
adenosine deaminase domain containing 1 (testis specific)
Gene Aliases
Tenr
Chromosome Location
Chr. 3: 37063093 - 37121930 on Build GRCm38
UniGene ID
Mm.8122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009350.3 NP_033376.2 2, 3 363
GenBank mRNA BC107346.2 - 2, 3 341
X84693.1 - 2, 3 351

View Full Product Details