Gene Transcripts

Gene Symbol
Tesk1
Entrez Gene ID
21754
Gene Name
testis specific protein kinase 1
Gene Aliases
AI326901
Chromosome Location
Chr. 4: 43442277 - 43448075 on Build GRCm38
UniGene ID
Mm.485441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011571.3 NP_035701.3 8 1958
GenBank mRNA AB003494.1 - 8 2296
AK144302.1 - 8 1541
BC099699.1 - 8 1958 6821934

View Full Product Details