Gene Transcripts
- Gene Symbol
- Tnfaip1
- Entrez Gene ID
- 21927
- Gene Name
- tumor necrosis factor, alpha-induced protein 1 (endothelial)
- Gene Aliases
- Edp-1, Edp1, Tnfip1
- Chromosome Location
- Chr. 11: 78522850 - 78536260 on Build GRCm38
- UniGene ID
- Mm.386774
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK034305.1 |
- |
3 |
529 |
|
View Full Product Details