Gene Transcripts

Gene Symbol
Tpo
Entrez Gene ID
22018
Gene Name
thyroid peroxidase
Gene Aliases
Chromosome Location
Chr. 12: 30054661 - 30132984 on Build GRCm38
UniGene ID
Mm.4991
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009417.2 NP_033443.1 5 425
GenBank mRNA AK047843.1 - 5 404
BC099421.1 - 4 357
X60703.1 - 5 425

View Full Product Details