Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009423.4 | NP_033449.2 | 5 | 714 | |
GenBank mRNA | AF233449.1 | - | 5 | 613 | |
AK035056.1 | - | 5 | 692 | ||
AK040877.1 | - | 692 | |||
AK046015.1 | - | 5 | 754 | ||
AK053900.1 | - | 5 | 711 | ||
BC020455.1 | - | 5 | 699 | ||
X92346.1 | - | 5 | 734 |