Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022813.3 | NP_073724.1 | 8 | 863 | |
GenBank mRNA | AF295402.1 | - | 8 | 867 | |
AK003080.1 | - | 8 | 863 | ||
AK161002.1 | - | 8 | 863 | ||
AK171766.1 | - | 8 | 854 | ||
BC003452.1 | - | 8 | 837 | ||
BC014751.1 | - | 8 | 840 | 4235556 |