Gene Transcripts
- Gene Symbol
- Ppnr
- Entrez Gene ID
- 26930
- Gene Name
- per-pentamer repeat gene
- Gene Aliases
- mp41
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.439757
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
S76755.1 |
- |
1 |
817 |
|
X65588.1 |
- |
1 |
817 |
|
View Full Product Details