Gene Transcripts

Gene Symbol
Ip6k1
Entrez Gene ID
27399
Gene Name
inositol hexaphosphate kinase 1
Gene Aliases
1200016D08Rik, AU022776, Ihpk1, InsP6, InsP6k1, Itpk6
Chromosome Location
Chr. 9: 108002648 - 108048782 on Build GRCm38
UniGene ID
Mm.276155
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013785.2 NP_038813.2 3 571
GenBank mRNA AF177144.1 - 2 271
AK088872.1 - 3 588
AK166143.1 - 3 575
AK171310.1 - 3 576
AK172921.1 - 3 630
AK180416.1 - 2 88
AK184546.1 - 2 88
AK202332.1 - 2 112
BC053030.1 - 3 576
BC059006.1 - 3 571 6827203

View Full Product Details