Gene Transcripts
- Gene Symbol
- H2afz
- Entrez Gene ID
- 51788
- Gene Name
- H2A histone family, member Z
- Gene Aliases
- H2A.Z, H2a.z-1
- Chromosome Location
- Chr. 3: 137864487 - 137866922 on Build GRCm38
- UniGene ID
- Mm.117541
- Species
- Mus musculus
- Gene Symbol
- Gm8203
- Entrez Gene ID
- 666634
- Gene Name
- predicted pseudogene 8203
- Gene Aliases
- EG666634
- Chromosome Location
- Chr. 6: 116173619 - 116174514 on Build GRCm38
- UniGene ID
- Mm.465508
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK210116.1 |
- |
1 |
309 |
|
View Full Product Details