Gene Transcripts

Gene Symbol
Sgta
Entrez Gene ID
52551
Gene Name
small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha
Gene Aliases
5330427H01Rik, AI194281, D10Ertd190e, Sgt, Stg
Chromosome Location
Chr. 10: 81044071 - 81060232 on Build GRCm38
UniGene ID
Mm.30068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024499.1 NP_078775.1 12 1082
GenBank mRNA AK039966.1 - 11 1077
AK051005.1 - 12 1081
AK079168.1 - 12 1092
AK165557.1 - 12 1106
AK183506.1 - 1 43
AK214284.1 - 1 43
BC003836.1 - 12 1082 3486148

View Full Product Details