Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016783.3 | NP_058063.2 | 2 | 450 | |
GenBank mRNA | AF042491.1 | - | 2 | 401 | |
AK153272.1 | - | 2 | 467 | ||
AK159101.1 | - | 2 | 467 | ||
AK159345.1 | - | 2 | 467 | ||
AK159599.1 | - | 2 | 467 | ||
AK162315.1 | - | 2 | 470 | ||
AK166240.1 | - | 2 | 467 | ||
AK167655.1 | - | 2 | 467 | ||
AK168387.1 | - | 2 | 466 | ||
AK168882.1 | - | 2 | 467 | ||
AK169063.1 | - | 3 | 410 | ||
AK193145.1 | - | 1 | 38 | ||
AK208626.1 | - | 1 | 38 | ||
AK214321.1 | - | 1 | 38 | ||
BC006016.1 | - | 2 | 450 | 3591536 |