Gene Transcripts
- Gene Symbol
- Sec61a1
- Entrez Gene ID
- 53421
- Gene Name
- Sec61 alpha 1 subunit (S. cerevisiae)
- Gene Aliases
- AA408394, AA410007, Sec61a
- Chromosome Location
- Chr. 6: 88503607 - 88518800 on Build GRCm38
- UniGene ID
- Mm.28375
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016906.4 |
NP_058602.1
|
4, 5 |
326 |
|
GenBank mRNA |
AB032902.1 |
- |
4, 5 |
338 |
|
AF145253.1 |
- |
4, 5 |
248 |
|
AF222743.1 |
- |
4, 5 |
248 |
|
AK088473.1 |
- |
4, 5 |
327 |
|
AK153055.1 |
- |
4, 5 |
316 |
|
AK159349.1 |
- |
4, 5 |
314 |
|
AK167484.1 |
- |
4, 5 |
256 |
|
AK219834.1 |
- |
3, 4 |
169 |
|
BC003707.1 |
- |
4, 5 |
252 |
3592772 |
View Full Product Details